Learn More
3M™ Formaldehyde Monitor 3271+
Formaldehyde Monitor 3721+ is a cost-effective and convenient way to perform personal air sampling for formaldehyde and certain other aldehydes.
Brand: 3M™ 7100251921
221.18 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- State of the Art Monitor Design
- Simple, cost-effective and convenient
- A valid method for air sampling and analysis of formaldehyde and certain other aldehydes
- Easy to use: Just clip the monitor to shirt lapel, collar, or pocket
- Unobtrusive: Monitors are small and lightweight and will not interfere with employee activities
- Documentation for industrial hygienists and IH labs available upon request
- Analysis is not included
Specifications
Formaldehyde | |
Personal Monitor | |
3721+ |
Product Content Correction
Product Title
3M™ Formaldehyde Monitor 3271+ > Detectable Analytes: Formaldehyde