missing translation for 'onlineSavingsMsg'
Learn More

FLASH Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB500-218MFV610

Product Code. 18770753

  • 548.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

FLASH Polyclonal antibody specifically detects FLASH in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FLASH
Polyclonal
Western Blot
CASP8 associated protein 2, caspase 8 associated protein 2, CED-4, FLASHKIAA1315CASP8-associated protein 2, FLICE associated huge, FLICE-associated huge protein, FLJ11208, human FLASH, RIP25FLASH homolog RIP25
N-terminus of the FLASH protein.
0.1 mL
Apoptosis, Cancer, Tumor Suppressors
9994
Store at 4C in the dark.
IgG
Western Blot
mFluor Violet 610 SE
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FLASH Antibody, mFluor Violet 610 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 610 SE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.