Learn More
Fisherbrand™ Urine Sedimentation System: Bulb Decanting Pipette
The Fisherbrand™ Urine Sedimentation System is designed to produce the same results as the most popular style urine sedimentation system using the same test procedure
Brand: Fisherbrand™ 14375560
This item is not returnable.
View return policy
Description
Decanting pipet allows supernatant to be decanted off; leaves 1mL of urine-containing sediment in the bottom of the tube.Specifications
Bulb Decanting Pipette | |
Designed to produce the same results as the most popular style Urine sedimentation system using the same test procedure | |
500 Case |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Urine Sedimentation System: Bulb Decanting Pipette