Learn More
Fisherbrand™ Hybridization Tubes
Fits FisherBiotech™ Hybridization Incubator rotisseries and most rotisseries that accept standard 35 or 70mm diameter tubes.
Brand:  Fisherbrand™ 1324770
 View more versions of this product
Product Code. 11791436
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Fits FisherBiotech™ Hybridization Incubator rotisseries and most rotisseries that accept standard 35 or 70mm diameter tubes.
  • Thick-walled borosilicate glass
  • Includes polypropylene cap
  • O-ring is available for 35mm tubes (sold separately, Cat. No. 13-247-RNG)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
70 mm
FisherBiotech hybridization incubator rotisseries
With Cap
Borosilicate Glass
2.75 in.
70 mm
11.81 in.
300 mm
Hybridization Tube