Learn More
Fisherbrand™ Disposable Base Molds
For use in the preparation of pathology specimens
Brand: Fisherbrand™ 22-363-553
This item is not returnable.
View return policy
Description
For use in the preparation of pathology specimens
- Available in common base mold sizes; can be used with the same styles and types of cassettes and embedding rings
- Molds are made from specially formulated plastic, offering excellent thermal exchange
- A smooth interior finish and round corners facilitate specimen removal
Specifications
Base Mold | |
15 mm | |
0.6 in. | |
0.5 cm | |
0.6 x 0.6 x 0.2 in. (15 x 15 x 5 mm) | |
Can be use with same styles and types of cassettes and embedding rings and provides offering excellent thermal exchange |
Plastic | |
15 mm | |
0.6 in. | |
0.19 in. | |
Single-use | |
500/Pk. |