Learn More
Fisherbrand™ Electrode Holder
Handles multiple pH and conductivity readings quickly with reduced electrode breakage. Fisherbrand Electrode Holder accepts all brands of standard electrodes; extends 21 in. and pivots 360°; designed for smooth movement and effortless operation.
Brand: Fisherbrand™ 90023-72
143.88 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Handles multiple pH and conductivity readings quickly with reduced electrode breakage. Fisherbrand Electrode Holder accepts all brands of standard electrodes; extends 21 in. and pivots 360°; designed for smooth movement and effortless operation.
- Articulated at three points, the arm holder pivots in any direction (360°) while holder maintains electrode in vertical position
- Extends to 21 in. (53cm) from weighted 8 in. (20cm), die-cast metal base
- Accommodates standard glass, reference, and combination electrodes as well as temperature and conductivity probes
- Spring counterbalance for smooth, fingertip control and stability
Specifications
Holder | |
pH |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%