missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Agar, Lennox (Granulated)

Used as a solidifying agent to provide the solid medium to support the growth of microbes

85.50€ - 326.00€

Specifications

Form Granulated Powder
Product Type Agar
For Use With (Application) Used as a solidifying agent to provide the solid medium to support the growth of microbes
View More Specs

Products 2
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
15400675
View Documents
Fisher BioReagents™
BP9745-2
2 kg Poly Pail This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15490665
View Documents
Fisher BioReagents™
BP9745-500
500 g Poly Bottle This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

  • Granulated LB Agar, Lennox media
  • CAS: [91079-40-2], [8013-01-2], [7647-14-5], [9002-18-0]
  • Store at room temperature

Specifications

Specifications

Granulated Powder
Agar
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • LB Agar, (Lennox L Agar), Granulated

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.