missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Agar, Lennox (Granulated)
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Brand: Fisher BioReagents™ BP9745-2
This item is not returnable.
View return policy
Description
- Granulated LB Agar, Lennox media
- CAS: [91079-40-2], [8013-01-2], [7647-14-5], [9002-18-0]
- Store at room temperature
Specifications
Granulated Powder | |
Used as a solidifying agent to provide the solid medium to support the growth of microbes | |
2 kg |
Agar | |
Poly Pail |
Safety and Handling
Product Identifier
- LB Agar, (Lennox L Agar), Granulated
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction