missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-21 Antibody (461804), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25372B
This item is not returnable.
View return policy
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
FGF-21 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
461804 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
FGF-21, fibroblast growth factor 21 | |
E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1 | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer | |
26291 | |
Store at 4°C in the dark. | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FGF-21 Antibody (461804), Biotin, Novus Biologicals™ > 0.1 mL; Biotin
Spot an opportunity for improvement?Share a Content Correction