Learn More
3M™ FFP1 Unvalved Particulate Respirator
3M™ FFP1 Unvalved Particulate Respirator provides FFP1 respiratory protection while adapting to the face shape for a comfortable fit.
Brand: 3M™ 7000062037
This item is not returnable.
View return policy
Description
- Provides FFP1 (NPF 4) protection against hazardous dusts and mists
- Classic cup shape respirators
- The lightweight, twin-strap design, nose foam, and nose clip help provide a comfortable, custom fit
- Features a durable, collapse-resistant inner shell
- Designed to work well with 3M™ Eyewear and Hearing Protection products
Specifications
CE Approved, EN 149: 2001 + A1: 2009 FFP1 NR D | |
White | |
Respirator | |
Cup |
Product Content Correction
Product Title
3M™ FFP1 Unvalved Particulate Respirator > Color: White