missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold
Superior for hybridoma cloning
Brand: MP Biomedicals™ 092916854
This item is not returnable.
View return policy
Description
- Collected from a USDA regulated and monitored abattoir by technicians who work “onsite”
- Aseptically collected via cardiac puncture
- Supplied sterile, with no detectable mycoplasma or viral infection

Specifications
USDA | |
-20°C | |
Fetal Bovine Serum | |
3.1 to 4.4 g/dL | |
Sterile | |
Used for cell culture and insect cell culture applications | |
27 mg/dL | |
280 to 325 mOsm/kg | |
USDA Inspected |
Pale to Golden Brown | |
Liquid | |
1 x 500 mL | |
Heat-inactivated | |
<1 ng/mL | |
Yes | |
No | |
6.7 to 7.8 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold > Fetal Bovine Serum, CellectGold
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction