Learn More
Thermo Scientific™ GeneRuler 100 bp Plus DNA Ladder
Shop All Thermo Scientific Products
Description
GeneRuler 100bp Plus DNA Ladder 100 to 3000bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 100bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye.
- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Bright reference bands (given in red)
- Supplied with loading dye for sample DNA
- Environmental benefits include:
- Recycled Content
Specifications
Specifications
Concentration | 6X |
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Orange G |
Product Type | DNA Ladder |
Ready to Load | No |
Size Range | 100 to 3000 bp |
Gel Compatibility | Agarose Gels |
Green Features | Sustainable packaging |
Product Line | GeneRuler |
Quantity | 50 μg |
Product Suggestions
Customers who viewed this item also viewed
Documents
Product Information
Manuals
Scientific Resources
Flyers
Posters
Reference Materials
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Buy 3 of the Same Product and Pay for 2
Limited-Time Offer on Essential Life Science ProductsPromo Code: 24113Expires: 2025-09-19
New Offer Save Now