Learn More
Greiner Bio-One F-Bottom 96-well Assay Microplates
Features a chimney-like arrangement of the wells that allows each well to stand on its own. Greiner Bio One™ F-Bottom 96-Well Assay Microplates are constructed of polystyrene, a highly clear polymer with excellent optical properties making it ideal for precise optical measurements. Polystyrene is also characterized by its ability to bind biomolecules, such as proteins and is suitable for work with cell cultures.
Brand: Greiner Bio-One 655948
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
- Manufactured from raw materials tested for leachables
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines

Specifications
DIN ISO 9001 | |
Standard | |
34 mm2 | |
Clear | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
-20°C to +60°C | |
Yes | |
Clear | |
5 mm |
RNase-/D-Nase, human DNA free | |
With Lid | |
8 x 12 | |
Polystyrene | |
25 to 34 μL | |
392 μL | |
Yes | |
4800 x g (swinging bucket rotor) | |
-20 to +60°C | |
11.5 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.