missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2/KMT6 Antibody (EZH2/6988), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20982PE
This item is not returnable.
View return policy
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Specifications
EZH2/KMT6 | |
Monoclonal | |
PE | |
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 | |
Recombinant fragment (around aa300-550) of human EZH2/KMT6 protein (exact sequence is proprietary) | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer | |
2146 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
EZH2/6988 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EZH2/KMT6 Antibody (EZH2/6988), PE, Novus Biologicals™ > 0.1 mL, PE
Spot an opportunity for improvement?Share a Content Correction