Learn More
Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications.
Brand: Applied Biosystems™ 75001.1.ML
467.40 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
- One-tube, one-step PCR cleanup
- Five-minute protocol
- Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
Specifications
-20°C | |
500 |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
7Hours
23Minutes
34Seconds
27