Learn More
Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions
Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.
Brand:  Thermo Scientific™ 01X211232
Product Code. 16455339
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Description:
  • ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
  • ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
  • ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number274-01 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog NumberECNACL45PPT,01X211232
Lot Number224-01 Certificate TypeCertificate of Analysis Date30/05/2024 Catalog NumberECNACL45PPT,01X211232
Lot Number462-01 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberECNACL45PPT,01X211232
Lot Number382-01 Certificate TypeCertificate of Analysis Date21/09/2022 Catalog NumberECNACL45PPT,01X211232
Lot Number242-01 Certificate TypeCertificate of Analysis Date15/06/2022 Catalog NumberECNACL45PPT,01X211232
Product Title
Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions > 45 ppt NaCl Calibration Solution