missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Specifications
Melting Point | 252°C |
Color | White |
pH | 4 to 6 |
Physical Form | Solid |
Quantity | 5 kg |
Formula Weight | 372.23g/mol |
Packaging | Plastic keg |
Safety and Handling


Product Identifier
- Ethylenediaminetetraacetic acid, disodium salt dihydrate
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction