Learn More
Ethyl 2-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals
68.70€ - 264.00€
Chemical Identifiers
CAS | 39574-16-8 |
---|---|
Molecular Formula | C6H9NO2S |
Molecular Weight (g/mol) | 159.20 |
MDL Number | MFCD00070636 |
InChI Key | ALJGYASQFZQQJX-UHFFFAOYNA-N |
Synonym | ethyl 2-isothiocyanatopropionate, ethyl n-thioxomethylene alaninate, propanoic acid, 2-isothiocyanato-, ethyl ester, acmc-20al0z, acmc-20lue7, propanoic acid, 2-isothiocyanato-, ethyl ester, s, ethyl l-2-isothiocyanatopropanoate, alanine, n-carbonothioyl-, ethyl ester |
PubChem CID | 142361 |
IUPAC Name | ethyl 2-isothiocyanatopropanoate |
SMILES | CCOC(=O)C(C)N=C=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11393216
|
Thermo Scientific Alfa Aesar
L12532.03 |
1 g |
68.70€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303226
|
Thermo Scientific Alfa Aesar
L12532.06 |
5 g |
264.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39574-16-8 | |
159.20 | |
ALJGYASQFZQQJX-UHFFFAOYNA-N | |
142361 | |
CCOC(=O)C(C)N=C=S |
C6H9NO2S | |
MFCD00070636 | |
ethyl 2-isothiocyanatopropionate, ethyl n-thioxomethylene alaninate, propanoic acid, 2-isothiocyanato-, ethyl ester, acmc-20al0z, acmc-20lue7, propanoic acid, 2-isothiocyanato-, ethyl ester, s, ethyl l-2-isothiocyanatopropanoate, alanine, n-carbonothioyl-, ethyl ester | |
ethyl 2-isothiocyanatopropanoate |
Specifications
39574-16-8 | |
C6H9NO2S | |
MFCD00070636 | |
1723243 | |
ethyl 2-isothiocyanatopropionate, ethyl n-thioxomethylene alaninate, propanoic acid, 2-isothiocyanato-, ethyl ester, acmc-20al0z, acmc-20lue7, propanoic acid, 2-isothiocyanato-, ethyl ester, s, ethyl l-2-isothiocyanatopropanoate, alanine, n-carbonothioyl-, ethyl ester | |
CCOC(=O)C(C)N=C=S | |
159.20 | |
159.2 | |
Ethyl 2-isothiocyanatopropionate |
63°C to 64°C (1 mmHg) | |
1.4925 | |
UN2922 | |
Moisture sensitive | |
ALJGYASQFZQQJX-UHFFFAOYNA-N | |
ethyl 2-isothiocyanatopropanoate | |
142361 | |
97% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H331-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only