Learn More
Ethyl 2-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals
Product Code. 11303226
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39574-16-8
Molecular Formula C6H9NO2S
Molecular Weight (g/mol) 159.20
MDL Number MFCD00070636
InChI Key ALJGYASQFZQQJX-UHFFFAOYNA-N
Synonym ethyl 2-isothiocyanatopropionate, ethyl n-thioxomethylene alaninate, propanoic acid, 2-isothiocyanato-, ethyl ester, acmc-20al0z, acmc-20lue7, propanoic acid, 2-isothiocyanato-, ethyl ester, s, ethyl l-2-isothiocyanatopropanoate, alanine, n-carbonothioyl-, ethyl ester
PubChem CID 142361
IUPAC Name ethyl 2-isothiocyanatopropanoate
SMILES CCOC(=O)C(C)N=C=S
Specifications
Boiling Point 63°C to 64°C (1 mmHg)
Refractive Index 1.4925
Quantity 5 g
UN Number UN2922
Beilstein 1723243
Sensitivity Moisture sensitive
Formula Weight 159.2
Percent Purity 97%
Chemical Name or Material Ethyl 2-isothiocyanatopropionate