missing translation for 'onlineSavingsMsg'
Learn More
Ethano L, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex(R), Thermo Scientific Chemicals Product Code.: 15883855

Ethano L, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex(R), Thermo Scientific Chemicals

Product Code. 15883855
2.5 L
Click to view available options
Quantity:
2.5 L
This item is not returnable. View return policy

Product Code. 15883855

Brand: Thermo Scientific Acros 463700025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114.1°C
Density 0.7900 g/mL
Boiling Point 78.0°C
Quantity 2.5 L
Flash Point 12°C
Assay Percent Range 95.0 % min. v/v
Specific Gravity 0.79
Linear Formula C2H5OH
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 46.06
Grade Extra Pure
Chemical Name or Material Ethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454665 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number463700025,463700050,463700250
Lot NumberA0455568 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number463700025,463700050,463700250
Lot NumberA0449788 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog Number463700025,463700050,463700250
Lot NumberA0448739 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number463700025,463700050,463700250
Lot NumberA0411875 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number463700025,463700050,463700250

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethano L, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex(R), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.