missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Equine TIMP-2 ELISA Kit
Equine TIMP-2 quantitates equine TIMP-2 in serum, plasma, supernatant.
678.00€ - 2635.00€
Specifications
Accession Number | collagenase inhibitor; CSC-21 K; D11Bwg1104e; DDC8; Metalloproteinase inhibitor 2; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; Timp2; TIMP-2; TIMP-2 protein; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2 |
---|---|
Assay Range | 16.38 to 4,000 pg/mL |
Assay Sensitivity | 4 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17144829
|
Invitrogen™
EE8RBX5 |
5 x 96 Tests |
2635.00€
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
17154363
|
Invitrogen™
EE8RB |
96 Tests |
678.00€
Pack of 96 |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
Description
Equine TIMP-2 quantitates equine TIMP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant equine TIMP-2.
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.Specifications
collagenase inhibitor; CSC-21 K; D11Bwg1104e; DDC8; Metalloproteinase inhibitor 2; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; Timp2; TIMP-2; TIMP-2 protein; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2 | |
4 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
TIMP2 | |
<10% | |
HRP | |
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL | |
TIMP2 | |
4 hr 45 min |
16.38 to 4,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O77717 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |