Learn More
Invitrogen™ Equine TIMP-2 ELISA Kit
Equine TIMP-2 quantitates equine TIMP-2 in serum, plasma, supernatant.
Brand:  Invitrogen™ EE8RB
 View more versions of this product
Product Code. 17154363
  • 678.00€ / Pack of 96
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Description
Equine TIMP-2 quantitates equine TIMP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant equine TIMP-2.
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
collagenase inhibitor; CSC-21 K; D11Bwg1104e; DDC8; Metalloproteinase inhibitor 2; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; Timp2; TIMP-2; TIMP-2 protein; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2
4 pg/mL
ELISA
Colorimetric Microplate Reader
TIMP2
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
16.38 to 4,000 pg/mL
Biotin
Plasma, Serum, Supernatant
O77717
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
TIMP2
4 hr 45 min