missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eotaxin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11786-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.Specifications
Eotaxin | |
Polyclonal | |
Unconjugated | |
CCL11 | |
C C motif chemokine 11, CCL11, Eosinophil chemotactic protein, Eotaxin, SCYA11, Small inducible cytokine A11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6356 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51671 | |
CCL11 | |
Eotaxin Fusion Protein Ag2398 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eotaxin Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction