missing translation for 'onlineSavingsMsg'
Learn More

Merck PureProteome™ Streptavidin Magnetic Bead System

Powerful bead system helps researchers to purify biotinylated molecules with high capacity, high specificity at an economical price

Brand:  Merck LSKMAGT02

Product Code. 12571219

  • 665.00€ / Each
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for immunoprecipitations, purifying nucleic acids, isolating cells and organelles
  • Binding capacity >20μg of protein per mg settled beads
  • High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Fast processing time: Beads are immobilized in seconds; increased kinetics of bead-protein binding enables shorter incubations
  • Particle Size: 10μm
  • Quantity: 2 x 1mL

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 x 1 mL
Store at 2° to 8°C.
Magnetic Beads
10 μm
Purification of Biotinylated molecules, Protein Purification
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions