missing translation for 'onlineSavingsMsg'
Learn More

Merck PureProteome™ Lambda Ig Binder Magnetic Beads

PureProteome™ magnetic beads are a powerful system to isolate antibodies faster, easier and with better reproducibly then ever before.

Brand:  Merck LSKMAGLM02

Product Code. 15905942

  • 830.00€ / Each
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket

Requires : PureProteome™ Magnetic Stand (8 well) (Cat # LSKMAGS08); Not delivered.
This item is not returnable. View return policy

Description

Description

The PureProteome™ Lambda Ig Binder Magnetic Bead System is a powerful system that helps researchers purify all Immunoglobulins (i.e IgG, IgA, IgM, IgE and IgD) with high specificity. Researchers can also purify/seperate the Fab portion from the Fc portion of Immunoglobulins.

  • High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Fast processing time: Beads are immobilized in seconds. Increased kinetics of bead-protein binding enables shorter incubations
  • Economical: Significantly more affordable then competitive magnetic beads

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Applications:

Antibody Purification

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lambda Ig binder Magnetic bead
10μm
Purifying all types of Igs Antibodies
2 x 1mL
2° to 8°C
Magnetic Beads
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions