missing translation for 'onlineSavingsMsg'
Learn More

Merck PureProteome™ Albumin Magnetic Beads

Faster, easier serum depletion with greater insight into your sample

Brand:  Merck LSKMAGL10

Product Code. 12521219

  • 972.00€ / 10mL
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Beads are conjugated to an antibody specific for human serum albumin
  • Provides rapid, scalable, and reproducible means to deplete >98% of albumin from serum and plasma samples, facilitating the detection and analysis of proteins of interest
  • Adjust bead volume based on sample volume and total protein concentration
  • No manipulation of tubes or multipart assemblies required
  • No need for centrifugation; magnetic field immobilizes beads in seconds
  • Particle Size: 10μm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Albumin Magnetic bead
10μm
Serum depletion
10mL
2° to 8°C
Magnetic Beads
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers