missing translation for 'onlineSavingsMsg'
Learn More
Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals Product Code.: 11472348

Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals

Product Code. 11472348
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11472348

Brand: Thermo Scientific Alfa Aesar A15722.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.

Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagiqueShow MoreShow Less
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Melting Point 450°C (decomposition)
Quantity 5 g
Beilstein 47549
Sensitivity Air and light sensitive
Merck Index 14,3547
Solubility Information Slightly soluble in water. Soluble in 1M sodium hydroxide (10mg/ml),EtOH,and pyridine.
Formula Weight 302.20 (Anhydrous)
Percent Purity 97%
Assay may cont. up to 12% water
Chemical Name or Material Ellagic acid hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240077 Certificate TypeCertificate of Analysis Date28/04/2022 Catalog NumberA15722.06,A15722.14,A15722.22
Lot Number10237783 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15722.06,A15722.14,A15722.22
Lot NumberJ9070B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15722.06,A15722.14,A15722.22
Lot Number10218793 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15722.06,A15722.14,A15722.22
Lot Number10172359 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15722.06,A15722.14,A15722.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ellagic acid hydrate, 97%, may cont. up to 12% water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.