missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EDTA Disodium Salt Dihydrate is a chelator of divalent cations. It inhibits enzymes, such as metalloproteases, that require divalent cations for activity. It is also used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Specifications
Specifications
Melting Point | 0°C |
Color | Colorless |
Density | 1g/cm3 |
pH | 4 to 5 |
Boiling Point | 100°C |
Odor | Odorless |
Quantity | 1 L |
Solubility Information | Soluble |
IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Formula Weight | 372.24g/mol |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EDTA Disodium Salt Solution, 0.01M, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction