missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Earplugs Zero Waste Box™
Recycle Earplugs using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
129.27€ - 280.15€
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17875755
|
Terracycle™
CS44-S-BE |
Small |
129.27€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
17815745
|
Terracycle™
CS44-M-BE |
Medium |
187.20€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
17815765
|
Terracycle™
CS44-L-BE |
Large |
280.15€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
Description
Use this Zero Waste Box to recycle:
- Foam and plastic earplugs
Approximate waste units accepted:
- Small: 880 earplugs
- Medium: 2400 earplugs
- Large: 4400 earplugs
Do not send in:
- Wax earplugs
- Hairnets, gloves, lab coats or other garments
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters and broken glass
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |