missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Earplugs Zero Waste Box™
GREENER_CHOICE

Recycle Earplugs using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Brand:  Terracycle™ CS44-M-BE

 View more versions of this product

Product Code. 17815745

  • 187.20€ / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use this Zero Waste Box to recycle:

  • Foam and plastic earplugs

Approximate waste units accepted:

  • Small: 880 earplugs
  • Medium: 2400 earplugs
  • Large: 4400 earplugs

Do not send in:

  • Wax earplugs
  • Hairnets, gloves, lab coats or other garments
  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters and broken glass
Specifications

Specifications

15 kg
Beige, Green, Red, White
Single-use
Medium
Not Autoclavable
Recycling Box
33 lb.
30.5 x 30.5 x 84 cm
Yes
Non-sterile
Cardboard, Plastic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions