Learn More
3M™ E-A-R™ Ultrafit™ Ear Plugs, Corded
Click to view available options
Color:
Green
Yellow
Yellow, Blue
Noise Reduction Rating (NRR):
14 db
20 db
32 db
Material:
Elastomeric Polymer
Silicone Rubber
TPE
Thermoplastic Elastomer
Quantity:
200/Pk.
50/Pk.
Description
- Triple-flange pliable plug effectively fits most ear canal sizes
- Reduces noise levels by 14-32 dB
- Ideal for dirty environments as no roll down required, simply push them into your ears for hygienic protection
- Clean easily with soapy water and reuse again and again, helping decrease waste
- Removable, reusable cord attachment – convenient if you move in and out of noisy environments
- Available with a storage case to keep plugs clean and together
Specifications
Specifications
Color | Yellow, Blue |
Certifications/Compliance | CE Approved |
Quantity | 50/Pk. |
Noise Reduction Rating (NRR) | 32 db |
Product Type | Ear Plugs |
Material | TPE |
Product Suggestions
Customers who viewed this item also viewed.
Documents
Product Information
Datasheets
7000103745
File size: 179.5 kB
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Ultrafit™ Ear Plugs, Corded >