Learn More
3M™ E-A-R™ Classic™ Earplugs
Click to view available options
Corded:
Corded
Noise Reduction Rating (NRR):
28db
Description
Made from a soft, energy-absorbing polymer foam that provides excellent hearing protection and all day comfort. Ideal for use where noise is a problem, either in work or leisure activities.
- Firm material with slow recovery time helps achieve good fit
- Special cylindrical shape that fits most ear canals and provides reliable seal
- Low equilibrium pressure helps reduce pressure in ear canal thus increasing comfort and wearability
- One size fits majority of wearers
- Excellent sound attenuation characteristics
- Sweat and humidity resistant which helps improve comfort
- Ideally suited for use in hot and humid conditions
Specifications
Specifications
Color | Yellow |
Certifications/Compliance | CE Approved |
Quantity | 200/Pk. |
Noise Reduction Rating (NRR) | 28db |
Corded | Corded |
Product Type | Classic Earplug |
Style | Roll-down |
Product Suggestions
Customers who viewed this item also viewed
Documents
Product Information
Datasheets
7000052844
File size: 413.6 kB
Promotions
-
Multiple Manufacturers
Outlet Corner
Use code 24095 to receive this promotion.Promo Code: 24095Expires: 2025-04-15
Days
12Hours
1Minutes
24Seconds
48 -
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Classic™ Earplugs >