Learn More
Abeba™ Dynamic 7335 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
71.20€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15652718
|
Abeba™
7335/35 |
35 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15662718
|
Abeba™
7335/36 |
36 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15672718
|
Abeba™
7335/37 |
37 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15682718
|
Abeba™
7335/38 |
38 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15692718
|
Abeba™
7335/39 |
39 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15602728
|
Abeba™
7335/40 |
40 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15612728
|
Abeba™
7335/41 |
41 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15622728
|
Abeba™
7335/42 |
42 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15632728
|
Abeba™
7335/43 |
43 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15642728
|
Abeba™
7335/44 |
44 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15652728
|
Abeba™
7335/45 |
45 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15662728
|
Abeba™
7335/46 |
46 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15672728
|
Abeba™
7335/47 |
47 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15682728
|
Abeba™
7335/48 |
48 |
71.20€
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber
- White/black
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Seamless upper
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Polyurethane |
White | |
Slip-resistant | |
7335 |