Learn More
Abeba™ Dynamic 7335 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7335/47
 View more versions of this product
Product Code. 15672728
  • 71.20€ / 1 pair
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber
  • White/black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Seamless upper
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
7335
White
47
Polyurethane