Learn More
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground
2-way tap plug with thread.
Brand: DWK Life Sciences 286036706
This item is not returnable.
View return policy
Description
Ground DURAN™ patent tap plugs have 2 almost parallel inclined bores.
- With thread, ground
- 2-way tap plug with 2 almost parallel inclined holes
- massive tap plug
Specifications
2.5 mm | |
Autoclavable | |
50/Pk. |
Borosilicate Glass (3.3) | |
14.5 NS | |
Installation in glass apparatus and devices |
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground > Bore 2.5 mm