Learn More
DWK Life Sciences DURAN™ Single way Stopcock, ground, without key
Ground shut-off taps without tap plug.
93.60€ - 735.00€
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | Coarse dosing of liquids; Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
15814778
|
DWK Life Sciences
286445403 |
18.8 NS | 4 mm |
344.00€
Pack of 25 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15834778
|
DWK Life Sciences
|
21.5 NS | 6 mm |
106.00€
Pack of 5 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15884768
|
DWK Life Sciences
286443805 |
14.5 NS (1.5 mm width) | 2.5 mm |
487.00€
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15854778
|
DWK Life Sciences
|
29.2 NS | 10 mm |
208.00€
Pack of 5 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15824778
|
DWK Life Sciences
286446005 |
18.8 NS | 6 mm |
406.00€
Pack of 25 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15874768
|
DWK Life Sciences
286443008 |
12.5 NS | 2 mm |
675.00€
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15864768
|
DWK Life Sciences
286442809 |
12.5 NS (1.5 mm width) | 2 mm |
449.00€
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15894768
|
DWK Life Sciences
|
14.5 NS | 2.5 mm |
735.00€
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15804778
|
DWK Life Sciences
286444407 |
14.5 NS | 4 mm |
569.00€
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15844778
|
DWK Life Sciences
286446802 |
24 NS | 8 mm |
164.00€
Pack of 5 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15854768
|
DWK Life Sciences
286442003 |
10 NS | 1.5 mm |
93.60€
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
Description
DURAN™ single way stopcocks are designed for use as shut-off valves. Valves are better suited for fine dosing. Ground DURAN™ single way stopcocks can be combined with - DURAN™ tap plug with groove - Threaded tap plug - hollow DURAN™ tap plugs
- Ground shut-off taps without tap plug
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Autoclavable | |
Coarse dosing of liquids; Installation in glass apparatus and devices |