Learn More
Lenz Laborglasinstrumente™ Duran Glass Columns acc. to Vigreux
60.00€ - 153.00€
Product Code | Brand | Length (Metric) | Includes | Standard Taper Joint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Includes | Standard Taper Joint | Price | Quantity & Availability | |||
10346261
|
Lenz Laborglasinstrumente™
05133301 |
300mm | With removable glass jacket | NS 14/23 |
124.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10396161
|
Lenz Laborglasinstrumente™
05130303 |
300mm | NS socket and NS cone | NS 24/29 |
98.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10747141
|
Lenz Laborglasinstrumente™
05130301 |
300mm | NS socket and NS cone | NS 14/23 |
80.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10181421
|
Lenz Laborglasinstrumente™
05130201 |
200mm | NS socket and NS cone | NS 14/23 |
60.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10234101
|
Lenz Laborglasinstrumente™
05130304 |
300mm | NS socket and NS cone | NS 29/32 |
91.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10355641
|
Lenz Laborglasinstrumente™
05133304 |
300mm | With removable glass jacket | NS 29/32 |
129.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10632282
|
Lenz Laborglasinstrumente™
05133504 |
600mm | With removable glass jacket | NS 29/32 |
153.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Specifications
Columns acc. to Vigreux | |
Duran Glass | |
1Each |
Product Title
Lenz Laborglasinstrumente™ Duran Glass Columns acc. to Vigreux