missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Duran™ Flat Flange Lids with Three Angled Side Neck and One Vertical Side Neck

362.00€ - 536.00€

Specifications

Material Borosilicate Glass, Duran Glass
For Use With (Equipment) Reaction Vessels
No. per Pack 1
Product Type Flat Flange Lid
Closure Type Flange Lid
View More Specs

Products 2
Product Code Brand Height (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Price Quantity & Availability  
10626433
View Documents
Lenz Laborglasinstrumente™
06105244
95mm
362.00€
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
10403455
View Documents
Lenz Laborglasinstrumente™
06105444
125mm
536.00€
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Borosilicate Glass, Duran Glass
1
Flange Lid
Reaction Vessels
Flat Flange Lid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ Duran™ Flat Flange Lids with Three Angled Side Neck and One Vertical Side Neck

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.