Learn More
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 03214158
This item is not returnable.
View return policy
Description
- Acc. to DIN 12394
- With side neck at 20° angle
- With inscription label
- Corrosion-free and resistant to thermal shock
- Ground joint features high taper accuracy and excellent tightness
Specifications
Acc. to DIN 12394 and in addition to DIN standard | |
Borosilicate Glass, Duran Glass | |
Round |
500 mL | |
14/23 NS Side Neck, 29/32 NS Center Neck |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled > Capacity/Socket Joint: 500mL-14/23 NS Side Neck, 29/32 NS Center Neck