missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (1D3), DyLight 405 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20111V
This item is not returnable.
View return policy
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
dsRNA | |
Monoclonal | |
DyLight 405 | |
Double-stranded RNA | |
RDV-RNA-methylated bovine serum albumin complex. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA, Immunohistochemistry | |
1D3 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
All species | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
dsRNA Antibody (1D3), DyLight 405 - Chimeric, Novus Biologicals™ > 0.1 mL; DyLight 405
Spot an opportunity for improvement?Share a Content Correction