missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (1D3), Alexa Fluor™ 700 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20110AF700
This item is not returnable.
View return policy
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
dsRNA | |
Monoclonal | |
Alexa Fluor 700 | |
Double-stranded RNA | |
RDV-RNA-methylated bovine serum albumin complex. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
1D3 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
dsRNA Antibody (1D3), Alexa Fluor™ 700 , Novus Biologicals™ > 0.1 mL; Alexa Fluor 700
Spot an opportunity for improvement?Share a Content Correction