Learn More
Thermo Scientific Chemicals Disposable IR cards, 5x10cm, KBr window, 15mm aperture
Cardboard card that fits into the holder of a spectrophotometer
Brand:  Thermo Scientific Chemicals 043936.KJ
125.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15446575
  • 125.63€ / Pack of 5
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Dimensiosn (L × H): 50 × 100mm (1.97 × 3.94 in.)
  • Disposable cardboard cards
  • Aperture: 15mm (0.591 in.)
  • KBr window
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IR Card
KBr window
5 Pk.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD26W033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number043937.KJ,043936.KM,043936.KJ
Lot NumberE06U017 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number043937.KJ,043936.KM,043936.KJ
Product Title
Thermo Scientific Chemicals Disposable IR cards, 5x10cm, KBr window, 15mm aperture > 5pc