missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylcarbazone-mercuric Reagent (I+II) EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
2 x 100 mL
Packaging:
Glass bottle
Specifications
Specifications
CAS | 6988-21-2 |
Packaging | Glass bottle |
Molecular Formula | C11H13NO4 |
Quantity | 2 x 100 mL |
Synonym | dioxacarb, dioxacarbe, dioxocarb, rovlinka, elocron, famid, elocron 50wp, mcdp, elocron 8353, du pont 1519 |
InChI Key | SDKQRNRRDYRQKY-UHFFFAOYSA-N |
SMILES | CNC(=O)OC1=CC=CC=C1C2OCCO2 |
IUPAC Name | [2-(1,3-dioxolan-2-yl)phenyl] N-methylcarbamate |
Molecular Weight (g/mol) | 223.228 |
PubChem CID | 23421 |
Show More |
Safety and Handling
Product Identifier
- Diphenylcarbazone mercuric reagent
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diphenylcarbazone-mercuric Reagent (I+II) EU Pharmacopoeia, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction