Learn More
Dimethylglyoxime, 99%, Thermo Scientific Chemicals
Description
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
Solubility
Soluble in alcohol, ether and solutions of alkalies. Insoluble in water.
Notes
Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | ∼240°C (decomposition) |
Quantity | 2500 g |
Beilstein | 506731 |
Merck Index | 14,3246 |
Solubility Information | Soluble in alcohol,ether and solutions of alkalies. Insoluble in water. |
Formula Weight | 116.12 |
Percent Purity | 99% |
Chemical Name or Material | Dimethylglyoxime |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62500084 | Certificate of Analysis | 12/02/2025 | A12810.22,A12810.36,A12810.0E |
62200060 | Certificate of Analysis | 30/01/2025 | A12810.22,A12810.36,A12810.0E |
Y25K001 | Certificate of Analysis | 26/11/2024 | A12810.22,A12810.36,A12810.0E |
U15E002 | Certificate of Analysis | 25/11/2024 | A12810.22,A12810.36,A12810.0E |
W24K056 | Certificate of Analysis | 03/10/2024 | A12810.22,A12810.36,A12810.0E |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.