missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals Product Code.: 11915791

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Product Code. 11915791
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11915791

Brand: Thermo Scientific Acros 116420050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 756-79-6
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.08
MDL Number MFCD00008349
InChI Key VONWDASPFIQPDY-UHFFFAOYSA-N
Synonym dimethyl methylphosphonate, dimethylmethylphosphonate, pyrol dmmp, dmmp, dimethyl methanephosphonate, fyrol dmmp, phosphonic acid, methyl-, dimethyl ester, methanephosphonic acid dimethyl ester, dimethoxymethylphosphine oxide, methylphosphonic acid dimethyl esterShow MoreShow Less
PubChem CID 12958
IUPAC Name [methoxy(methyl)phosphoryl]oxymethane
SMILES COP(C)(=O)OC

Specifications

Color Colorless
Density 1.1400g/mL
Boiling Point 181.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2P(O)CH3
Refractive Index 1.413
Quantity 5 g
Beilstein 04,I,572
Fieser 03,117; 11,203
Merck Index 15,3435
Specific Gravity 1.14
Formula Weight 124.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethyl methylphosphonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0412344 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number116420250,116420050
Lot NumberA0410494 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number116420250,116420050
Lot NumberA0374608 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number116420250,116420050
Lot NumberA0415604 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number116420250,116420050
Lot NumberA0448130 Certificate TypeCertificate of Analysis Date30/11/2022 Catalog Number116420250,116420050
compliance-icons compliance-icons
Product Identifier
  • Dimethyl methylphosphonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.