missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Hamilton Microlab 500™

Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.

82.65€ - 87.85€

Specifications

Description 60mm stroke, 6-32 UNC thread in plunger button
Includes Plunger stop
Length (Metric) 60 mm
Product Type Hamilton MicroLab 500™ Instrument Syringes
Connection Type Luer Lock
View More Specs

Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15298483
View Documents
Trajan Scientific and Medical™
006232
250 μL
87.55€
Each
Please to purchase this item. Need a web account? Register with us today!
15238483
View Documents
Trajan Scientific and Medical™
005232
100 μL
82.65€
Each
Please to purchase this item. Need a web account? Register with us today!
15268473
View Documents
Trajan Scientific and Medical™
004232
50 μL
87.85€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for Hamilton Microlab 500
Specifications

Specifications

60mm stroke, 6-32 UNC thread in plunger button
60 mm
Luer Lock
Plunger stop
Hamilton MicroLab 500™ Instrument Syringes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.