Learn More
Aalborg™ Digital Mass Flowmeter
Use a local four button teypad and LCD to access all features of the flow meter. Aalborg™ Digital Mass Flowmeter supports automatic sensor zero offset adjustments.
Brand:  Aalborg™ DFM26S-VAL2-AA5/3AB-03-SS
 View more versions of this product
Product Code. 11727629
  • 2315.00€ / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Multi-drop capability of up to 255 units (for RS-485 option)
  • Stores calibration data for up to ten different gases
  • Supports ten different engineering units including user defined
  • Programmable 12 digits totalizer indicates total gas volume
  • Flow alarm limits for high, low gas flow with relay output
  • Pressure Alarm limits for high, low gas pressure with relay output
  • Temperature alarm limits for high low gas temperature with relay output
  • Four button keypad large 128 x 64 graphical LCD with back light
  • Digital (RS-232 or RS-485), analog outputs operate simultaneously
  • Internal conversion factors for up to 32 gases
  • Automatic zero adjustment
  • Self-diagnostic tests
Certifications
NIST
Specifications
Specifications
Flowmeter
0 to 10 mL/min.
0.63 cm (0.25 in.) Standard Compression Fittings
Programs, records, analyze flow rates of various gases with computer via an RS-485 interface.
32° to 122°F
±1%
5 to 100 psig
Digital Mass Flowmeter
±0.15% (Full Scale)
0°C to 50°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products