Learn More
Mitutoyo™ Digimatic DMX-1 D-Sub9 Serial Interface Connector
Microcontrolled D-SUB9 cable serial interface for connecting a single measurement device via the RS-232C computer interface. Mitutoyo™ Digimatic DMX-1 D-SUB9 Serial Interface Connector requires no power supply.
Brand: Mitutoyo™ 011216
Description
- Microcontrolled interface
- Connects a single measurment device, with Digimatic output, to an RS-232C computer interface.
- Easily connects to a computer's RS-232C serial port.
- No external power supply is needed, as the small level of power needed is supplied by the handshake lines RTS and DTR.
Specifications
DMX-1 D-SUB9 Serial Interface | |
Computer/measurement device | |
62 mm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.