Learn More
Diethanolamine, ACS reagent, Thermo Scientific Chemicals Product Code.: 15276008
compliance-icons compliance-icons compliance-icons
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Product Code. 15276008
1 kg
Click to view available options
Quantity:
250 g
1 kg
Product Code. 15276008
Brand: Thermo Scientific Acros 449060010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobisShow MoreShow Less
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO
Specifications
Melting Point 27.0°C to 30.0°C
Density 1.0955g/mL
Boiling Point 268.0°C
Flash Point 179°C
Linear Formula (HOCH2CH2)2NH
Refractive Index 1.4750 to 1.4790
Quantity 1 kg
Merck Index 15,3118
Specific Gravity 1.0955
Solubility Information Solubility in water: 954g/L (20°C).
Viscosity 352 mPa.s (30°C)
Formula Weight 105.14
Percent Purity ≥98.5% (GC) 1% max. (2-Aminoethanol) (GC) 1.0% max. (Triethanolamine) (GC)
Grade ACS Reagent
Chemical Name or Material Diethanolamine
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468295 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number449060010,449062500
Lot NumberA0453742 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number449060010,449062500
Lot NumberA0390741 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number449060010,449062500
Lot NumberA0440805 Certificate TypeCertificate of Analysis Date08/04/2022 Catalog Number449060010,449062500
Lot NumberA0418331 Certificate TypeCertificate of Analysis Date11/10/2021 Catalog Number449060010,449062500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
RUO – Research Use Only
Product Content Correction
Product Title
Diethanolamine, ACS reagent, Thermo Scientific Chemicals >