missing translation for 'onlineSavingsMsg'
Learn More
Diaminomaleonitrile, 98%, Thermo Scientific Chemicals Product Code.: 10104290

Diaminomaleonitrile, 98%, Thermo Scientific Chemicals

Product Code. 10104290
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10104290

Brand: Thermo Scientific Acros 172701000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187-42-4
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.1
MDL Number MFCD00001870
InChI Key DPZSNGJNFHWQDC-ARJAWSKDSA-N
Synonym diaminomaleonitrile, 2,3-diaminomaleonitrile, hydrogen cyanide tetramer, maleonitrile, diamino, ccris 921, 2-butenedinitrile, 2,3-diamino-, 2z, 2z-diaminobut-2-enedinitrile, 2,3-diamino maleonitrile, 2-butenedinitrile, 2,3-diamino-, z, z-2,3-diaminobut-2-enedinitrile
PubChem CID 2723951
IUPAC Name (Z)-2,3-diaminobut-2-enedinitrile
SMILES C(#N)C(=C(C#N)N)N

Specifications

Melting Point 178.0°C to 179.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NCC(NH2)=C(NH2)CN
Quantity 100 g
Beilstein 04, II, 949
Fieser 05,175; 04,480; 13,327; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Solubility Information Solubility in water: 5g/L (25°C). Other solubilities: soluble in methanol, ethanol, dmf and dimethyl, sulfoxide, slightly soluble in ethyl ether and dioxane, insol. in benzene,acetone,xylene,toluene and cs2
Formula Weight 108.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diaminomaleonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443056 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number172701000,172700250,172705000
Lot NumberA0414015 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number172701000,172700250,172705000
Lot NumberA0387807 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number172701000,172700250,172705000
Lot NumberA0444834 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog Number172701000,172700250,172705000
Lot NumberA0443056 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number172701000,172700250,172705000
compliance-icons
Product Identifier
  • Diaminomaleonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diaminomaleonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.